Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012797.2 | NP_036929.2 | 2 | 842 | ||
XM_006235267.2 | XP_006235329.1 | 2 | 1813 | |||
GenBank mRNA | BC127448.1 | - | 2 | 854 | 8372348 | |
L23148.1 | - | 1 | 1064 | |||
M86708.1 | - | 2 | 849 |