Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012808.1 | NP_036940.1 | 1 | 515 | ||
GenBank mRNA | BC088449.1 | - | 1 | 515 | 7126617 | |
X56228.1 | - | 1 | 478 |