Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135585.1 | NP_001129057.1 | 3 | 450 | ||
NM_003039.2 | NP_003030.1 | 3 | 450 | |||
XM_005263491.3 | XP_005263548.1 | 4 | 248 | |||
XM_017002133.1 | XP_016857622.1 | 5 | 476 | |||
XM_017002134.1 | XP_016857623.1 | 5 | 442 | |||
XM_017002135.1 | XP_016857624.1 | 5 | 536 | |||
XM_017002136.1 | XP_016857625.1 | 5 | 512 | |||
XM_017002137.1 | XP_016857626.1 | 6 | 548 | |||
XM_017002138.1 | XP_016857627.1 | 6 | 524 | |||
XM_017002139.1 | XP_016857628.1 | 4 | 458 | |||
XM_017002140.1 | XP_016857629.1 | 5 | 379 | |||
XM_017002141.1 | XP_016857630.1 | 4 | 281 | |||
GenBank mRNA | AK289849.1 | - | 3 | 206 | ||
AK290398.1 | - | 3 | 240 | |||
AK295785.1 | - | 3 | 218 | |||
AK302047.1 | - | 3 | 91 | |||
AK302057.1 | - | 6 | 374 | |||
AK302137.1 | - | 5 | 425 | |||
BC001692.2 | - | 3 | 209 | 2966459 | ||
BC001820.2 | - | 3 | 209 | 2966459 | ||
BC035878.1 | - | 3 | 237 | |||
DC379829.1 | - | 3 | 450 | |||
M55531.1 | - | 3 | 233 |