Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000341.3 | NP_000332.2 | 9, 10 | 1690 | ||
GenBank mRNA | AB033549.1 | - | 9, 10 | 1671 | ||
AK091932.1 | - | 10, 11 | 1556 | |||
AK092000.1 | - | 6, 7 | 1796 | |||
AK223146.1 | - | 9, 10 | 1652 | |||
AK289636.1 | - | 9, 10 | 1668 | |||
AK313157.1 | - | 9, 10 | 1684 | |||
BC022386.1 | - | 9, 10 | 1688 | 4556946 | ||
BC093624.1 | - | 9, 10 | 1649 | 7939469 | ||
BC093626.1 | - | 9, 10 | 1644 | 7939471 | ||
DQ023512.1 | - | 6, 7 | 1796 | |||
DQ023514.1 | - | 3, 4 | 813 | |||
KU178335.1 | - | 9, 10 | 1612 | |||
L11696.1 | - | 9, 10 | 1655 | |||
M95548.1 | - | 9, 10 | 1656 |