Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cebpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012831.1 | NP_036963.1 | 2 | 528 | ||
XM_008759112.1 | XP_008757334.1 | 3 | 771 | |||
GenBank mRNA | BC166589.1 | - | 2 | 517 | 7934938 | |
X64403.1 | - | 2 | 528 |