Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012832.3 | NP_036964.3 | 4 | 337 | ||
XM_008759497.1 | XP_008757719.1 | 2 | 4195 | |||
XM_008759498.1 | XP_008757720.1 | 4 | 397 | |||
GenBank mRNA | AY574256.1 | - | 4 | 337 | ||
AY671974.1 | - | 4 | 340 | |||
L31619.1 | - | 4 | 359 | |||
S53987.1 | - | 378 |