Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cycs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012839.2 | NP_036971.1 | 3 | 1154 | ||
GenBank mRNA | BC081849.1 | - | 3 | 1154 | 7192255 | |
BC089051.1 | - | 3 | 1165 | 7305562 | ||
FQ211631.1 | - | 3 | 1146 | |||
FQ212556.1 | - | 3 | 1148 | |||
FQ213464.1 | - | 3 | 1142 | |||
FQ214733.1 | - | 3 | 1146 | |||
FQ214738.1 | - | 3 | 1144 | |||
FQ214908.1 | - | 3 | 1146 | |||
FQ215052.1 | - | 3 | 1146 | |||
FQ215466.1 | - | 3 | 1146 | |||
FQ215858.1 | - | 3 | 1189 | |||
FQ216163.1 | - | 3 | 1148 | |||
FQ216426.1 | - | 3 | 1147 | |||
FQ216512.1 | - | 3 | 1146 | |||
FQ216560.1 | - | 3 | 1147 | |||
FQ216674.1 | - | 3 | 1145 | |||
FQ216679.1 | - | 3 | 1146 | |||
FQ217283.1 | - | 3 | 1146 | |||
FQ217634.1 | - | 3 | 1146 | |||
FQ218428.1 | - | 3 | 1146 | |||
FQ218668.1 | - | 3 | 1147 | |||
FQ220222.1 | - | 3 | 1177 | |||
FQ223761.1 | - | 3 | 1146 | |||
FQ224710.1 | - | 3 | 1146 | |||
FQ224914.1 | - | 3 | 1189 | |||
FQ224919.1 | - | 3 | 1147 |
Gene Symbol: LOC100363502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751799.3 | XP_003751847.1 | 1 | 1150 |