Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003041.3 | NP_003032.1 | 1 | 90 | ||
NR_130783.1 | 1 | 90 | ||||
XM_006721072.3 | XP_006721135.2 | 1 | 673 | |||
GenBank mRNA | BC131542.1 | - | 1 | 90 | 40085355 | |
M95549.1 | - | 1 | 91 |