Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMTN SIRNA Assays ›
Gene Symbol:
SMTN
Gene Name:
smoothelin
Gene Aliases:
-
Chromosome Location:
Chr.22: 31081296 - 31104624 on Build GRCh38
UniGene ID:
Hs.149098
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12959
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001207017.1
NP_001193946.1
14
2161
NM_001207018.1
NP_001193947.1
12
2090
NM_006932.4
NP_008863.3
13
2086
NM_134269.2
NP_599031.1
13
2086
NM_134270.2
NP_599032.2
13
2086
XM_006724298.1
XP_006724361.1
14
2157
XM_006724302.1
XP_006724365.1
14
2216
XM_011530333.2
XP_011528635.1
15
2262
XM_011530336.2
XP_011528638.1
15
2263
XM_011530337.2
XP_011528639.1
15
2263
XM_011530338.2
XP_011528640.1
14
2169
XM_011530339.2
XP_011528641.1
15
2263
XM_011530340.2
XP_011528642.1
14
2170
XM_011530341.1
XP_011528643.1
7
1094
XM_017028913.1
XP_016884402.1
15
2263
XM_017028914.1
XP_016884403.1
14
2169
XM_017028915.1
XP_016884404.1
14
2169
XM_017028916.1
XP_016884405.1
15
2263
XM_017028917.1
XP_016884406.1
15
2263
XM_017028918.1
XP_016884407.1
15
2263
XM_017028919.1
XP_016884408.1
14
2169
GenBank mRNA
AB209643.1
-
9
1737
AF064238.3
-
13
2063
AJ010306.2
-
13
2063
AK092684.1
-
3
732
AK095916.1
-
4
505
AK095930.1
-
4
618
AK096953.1
-
13
2121
AK128627.1
-
13
3724
AK128719.1
-
13
1932
AK225648.1
-
4
527
AK293681.1
-
4
460
AK295418.1
-
12
2090
AK300064.1
-
4
607
AK301289.1
-
3
362
AK304084.1
-
14
2161
AK307627.1
-
4
535
AK309502.1
-
4
599
AY061971.1
-
13
2064
AY061972.1
-
13
2064
BC034237.2
-
13
2054
3912887
Y13492.2
-
13
2064
Z49989.2
-
4
520
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
smooth muscle contraction
muscle organ development
actin binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbzcs:80/10.239.166.68:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline