Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003042.3 | NP_003033.3 | 5 | 802 | ||
XM_005265410.4 | XP_005265467.1 | 5 | 2159 | |||
XM_005265411.4 | XP_005265468.1 | 5 | 2098 | |||
XM_006713306.3 | XP_006713369.1 | 5 | 757 | |||
XM_011534025.2 | XP_011532327.1 | 5 | 2849 | |||
XM_011534027.2 | XP_011532329.1 | 5 | 2788 | |||
XM_017007071.1 | XP_016862560.1 | 6 | 2581 | |||
XM_017007072.1 | XP_016862561.1 | 6 | 1891 | |||
GenBank mRNA | AK295702.1 | - | 2 | 165 | ||
BC033904.1 | - | 5 | 770 | 5264672 | ||
BC057291.1 | - | 5 | 782 | |||
X54673.1 | - | 4 | 625 |