Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033715.1 | NP_001028887.1 | 23 | 1912 | ||
NM_001033716.1 | NP_001028888.1 | 22 | 1843 | |||
NM_053295.2 | NP_445747.2 | 24 | 2026 | |||
XM_006231698.2 | XP_006231760.1 | 24 | 2434 | |||
XM_006231699.2 | XP_006231761.1 | 23 | 2159 | |||
GenBank mRNA | AF346597.1 | - | 20 | 1524 | ||
AF346598.1 | - | 22 | 1707 | |||
BC091239.1 | - | 23 | 1912 | 7324026 | ||
DQ186624.1 | - | 24 | 2220 | |||
DQ186625.1 | - | 22 | 2037 | |||
DQ186626.1 | - | 23 | 2151 | |||
DQ186627.1 | - | 23 | 2106 | |||
DQ186628.1 | - | 22 | 2067 | |||
DQ186629.1 | - | 21 | 1998 | |||
DQ287975.1 | - | 17 | 1898 | |||
Y13587.1 | - | 21 | 1605 | |||
Y13588.1 | - | 22 | 1719 | |||
Y13589.1 | - | 20 | 1536 |