Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CK357656.1 | - | 1 | 190 | ||
FQ217150.1 | - | 1 | 220 | |||
FQ217655.1 | - | 1 | 222 | |||
FQ220032.1 | - | 479 | ||||
FQ221038.1 | - | 1 | 219 | |||
FQ221154.1 | - | 1 | 210 | |||
FQ221373.1 | - | 1 | 220 | |||
FQ222277.1 | - | 1 | 133 | |||
FQ222316.1 | - | 1 | 219 | |||
FQ222356.1 | - | 1 | 220 | |||
FQ222897.1 | - | 1 | 221 | |||
FQ223053.1 | - | 1 | 219 | |||
FQ224290.1 | - | 1 | 221 | |||
FQ224782.1 | - | 1 | 220 | |||
FQ228448.1 | - | 1 | 219 | |||
FQ228589.1 | - | 1 | 220 | |||
M55534.1 | - | 759 | ||||
S74229.1 | - | 1 | 183 | |||
S77138.1 | - | 1 | 197 | |||
S77142.1 | - | 1 | 182 | |||
X60351.1 | - | 1 | 189 |