Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CK357656.1 | - | 3 | 500 | ||
FQ217150.1 | - | 3 | 530 | |||
FQ217655.1 | - | 3 | 532 | |||
FQ220032.1 | - | 789 | ||||
FQ220162.1 | - | 3 | 531 | |||
FQ221038.1 | - | 3 | 529 | |||
FQ221154.1 | - | 3 | 520 | |||
FQ221373.1 | - | 3 | 530 | |||
FQ222161.1 | - | 3 | 530 | |||
FQ222277.1 | - | 3 | 443 | |||
FQ222316.1 | - | 3 | 529 | |||
FQ222356.1 | - | 3 | 530 | |||
FQ222897.1 | - | 3 | 531 | |||
FQ223053.1 | - | 3 | 529 | |||
FQ224290.1 | - | 3 | 531 | |||
FQ224782.1 | - | 3 | 530 | |||
FQ228448.1 | - | 3 | 529 | |||
FQ228589.1 | - | 3 | 530 | |||
M24092.1 | - | 1 | 71 | |||
M55534.1 | - | 1069 | ||||
S74229.1 | - | 3 | 493 | |||
S77138.1 | - | 3 | 507 | |||
S77142.1 | - | 3 | 492 | |||
X60351.1 | - | 3 | 499 |