Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012948.1 | NP_037080.1 | 3, 4 | 381 | ||
GenBank mRNA | BC059151.1 | - | 3, 4 | 394 | 6919042 | |
X98377.1 | - | 3, 4 | 381 |