Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012949.2 | NP_037081.2 | 8 | 812 | ||
XM_006246599.2 | XP_006246661.1 | 8 | 883 | |||
XM_006246600.2 | XP_006246662.1 | 8 | 863 | |||
GenBank mRNA | BC083566.1 | - | 8 | 799 | 7193009 | |
FQ214810.1 | - | 7 | 790 | |||
FQ214945.1 | - | 8 | 830 | |||
FQ214984.1 | - | 8 | 823 | |||
FQ215034.1 | - | 4 | 489 | |||
FQ215221.1 | - | 7 | 724 | |||
FQ215310.1 | - | 4 | 490 | |||
FQ215357.1 | - | 8 | 834 | |||
FQ215526.1 | - | 8 | 822 | |||
FQ215607.1 | - | 4 | 489 | |||
FQ215648.1 | - | 8 | 822 | |||
FQ215780.1 | - | 4 | 489 | |||
FQ215843.1 | - | 7 | 790 | |||
FQ215865.1 | - | 8 | 822 | |||
FQ216150.1 | - | 7 | 791 | |||
FQ216271.1 | - | 4 | 490 | |||
FQ216382.1 | - | 8 | 825 | |||
FQ216386.1 | - | 7 | 800 | |||
FQ216660.1 | - | 7 | 790 | |||
FQ216781.1 | - | 4 | 489 | |||
FQ217095.1 | - | 8 | 822 | |||
FQ223831.1 | - | 4 | 489 | |||
Y00979.1 | - | 7 | 774 |