Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012949.2 | NP_037081.2 | 3 | 203 | ||
XM_006246599.2 | XP_006246661.1 | 3 | 274 | |||
XM_006246600.2 | XP_006246662.1 | 3 | 254 | |||
GenBank mRNA | BC083566.1 | - | 3 | 190 | 7193009 | |
DY321069.1 | - | 3 | 203 | |||
FQ214810.1 | - | 2 | 181 | |||
FQ214945.1 | - | 3 | 221 | |||
FQ214984.1 | - | 3 | 213 | |||
FQ215221.1 | - | 2 | 116 | |||
FQ215357.1 | - | 3 | 225 | |||
FQ215648.1 | - | 3 | 213 | |||
FQ215759.1 | - | 3 | 225 | |||
FQ215843.1 | - | 2 | 180 | |||
FQ215865.1 | - | 3 | 213 | |||
FQ216150.1 | - | 2 | 181 | |||
FQ216382.1 | - | 3 | 213 | |||
FQ216386.1 | - | 2 | 190 | |||
FQ216660.1 | - | 2 | 180 | |||
FQ217095.1 | - | 3 | 213 | |||
FQ224938.1 | - | 3 | 213 | |||
Y00979.1 | - | 2 | 165 |