Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fosl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012953.1 | NP_037085.1 | 3 | 616 | ||
GenBank mRNA | BC097342.1 | - | 3 | 595 | 7459558 | |
M19651.1 | - | 3 | 616 |