Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014228.4 | NP_055043.2 | 10 | 1689 | ||
XM_017009767.1 | XP_016865256.1 | 11 | 1596 | |||
XM_017009768.1 | XP_016865257.1 | 11 | 1672 | |||
XM_017009769.1 | XP_016865258.1 | 11 | 3203 | |||
XM_017009770.1 | XP_016865259.1 | 11 | 1323 | |||
GenBank mRNA | AK096607.1 | - | 10 | 1593 | ||
AK308895.1 | - | 10 | 1465 | |||
AK312309.1 | - | 10 | 1469 | |||
BC069631.1 | - | 10 | 1222 | 7262437 | ||
BC093785.1 | - | 10 | 1298 | 7939630 | ||
BC113425.1 | - | 10 | 1298 | 8322477 | ||
S80071.1 | - | 10 | 1222 |