Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012976.2 | NP_037108.1 | 2 | 121 | ||
GenBank mRNA | FQ225275.1 | - | 2 | 115 | ||
FQ225984.1 | - | 2 | 137 | |||
FQ226981.1 | - | 2 | 142 | |||
FQ227276.1 | - | 2 | 136 | |||
FQ228081.1 | - | 2 | 141 | |||
FQ230182.1 | - | 2 | 141 | |||
FQ231538.1 | - | 2 | 140 | |||
FQ231564.1 | - | 2 | 151 | |||
FQ232247.1 | - | 2 | 162 | |||
FQ232513.1 | - | 2 | 144 | |||
FQ233614.1 | - | 2 | 162 | |||
FQ234601.1 | - | 2 | 144 | |||
FQ234845.1 | - | 2 | 164 | |||
L21711.1 | - | 2 | 130 | |||
L36862.1 | - | 2 | 121 |