Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001632.4 | NP_001623.3 | 11 | 2599 | ||
GenBank mRNA | AF217992.1 | - | 1 | 756 | ||
AK075432.1 | - | 11 | 2414 | |||
BC009647.1 | - | 11 | 2396 | 3460735 | ||
BC068501.1 | - | 11 | 2382 | 30349481 | ||
BC094743.1 | - | 11 | 2462 | 30330997 | ||
M12551.1 | - | 11 | 2339 | |||
M14169.1 | - | 10 | 2279 | |||
M14170.1 | - | 11 | 2343 |