Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138878.2 | NP_620233.1 | 2 | 303 | ||
GenBank mRNA | AF095740.1 | - | 2 | 102 | ||
BC084728.1 | - | 2 | 123 | 7315541 | ||
CB581170.1 | - | 2 | 314 | |||
FQ230524.1 | - | 1 | 29 |