Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105720.2 | NP_001099190.2 | 2 | 377 | ||
GenBank mRNA | BC166886.1 | - | 2 | 329 | 6890357 | |
CB316505.1 | - | 2 | 331 | |||
FQ215246.1 | - | 2 | 329 | |||
FQ225350.1 | - | 2 | 324 | |||
FQ225877.1 | - | 2 | 327 | |||
FQ226288.1 | - | 2 | 327 | |||
FQ227283.1 | - | 2 | 327 | |||
FQ233200.1 | - | 2 | 327 | |||
FQ233557.1 | - | 2 | 329 | |||
FQ233627.1 | - | 2 | 315 | |||
FQ234262.1 | - | 2 | 329 | |||
X63594.1 | - | 2 | 307 |