Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142367.2 | NP_001135839.1 | 1, 2 | 354 | ||
NM_001285415.1 | NP_001272344.1 | 1, 2 | 354 | |||
NM_012997.3 | NP_037129.1 | 1, 2 | 354 | |||
GenBank mRNA | AF231010.1 | - | 1, 2 | 145 | ||
BC061742.1 | - | 1, 2 | 346 | 5599053 | ||
FJ424511.1 | - | 1, 2 | 145 | |||
X80477.1 | - | 1, 2 | 354 |