Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318900.1 | NP_001305829.1 | 13 | 2611 | ||
NM_001318901.1 | NP_001305830.1 | 16 | 2964 | |||
NM_001318902.1 | NP_001305831.1 | 15 | 2891 | |||
NM_001318904.1 | NP_001305833.1 | 14 | 3406 | |||
NM_001318905.1 | NP_001305834.1 | 16 | 3532 | |||
NM_001318906.1 | NP_001305835.1 | 14 | 3239 | |||
NM_005271.4 | NP_005262.1 | 13 | 3179 | |||
GenBank mRNA | AB209406.1 | - | 12 | 5479 | ||
AF086070.1 | - | 1 | 87 | |||
AK094782.1 | - | 15 | 2843 | |||
AK122685.1 | - | 16 | 2948 | |||
BC112946.1 | - | 13 | 2962 | 6138490 | ||
M20867.1 | - | 13 | 2999 | |||
M37154.1 | - | 13 | 2913 | |||
X07674.1 | - | 13 | 2913 | |||
X07769.1 | - | 13 | 2913 |