Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013015.2 | NP_037147.1 | 5 | 522 | ||
GenBank mRNA | FQ214359.1 | - | 4 | 774 | ||
J04488.1 | - | 5 | 522 |