Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013019.2 | NP_037151.2 | 5 | 429 | ||
XM_008772599.1 | XP_008770821.1 | 5 | 1268 | |||
GenBank mRNA | BC062016.1 | - | 5 | 429 | 5621494 | |
X06890.1 | - | 5 | 361 |