Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138708.2 | NP_619715.1 | 5, 6 | 599 | ||
GenBank mRNA | BC060536.1 | - | 623 | |||
BC078683.1 | - | 5, 6 | 588 | 7135106 | ||
FQ214773.1 | - | 603 | ||||
FQ216744.1 | - | 603 | ||||
FQ224712.1 | - | 602 | ||||
FQ229109.1 | - | 603 | ||||
L10416.1 | - | 5, 6 | 599 | |||
S62097.1 | - | 5, 6 | 599 |