Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008539.3 | NP_001008539.3 | 4 | 736 | ||
NM_001164771.1 | NP_001158243.1 | 4 | 732 | |||
NM_003046.5 | NP_003037.4 | 4 | 732 | |||
XM_005273610.4 | XP_005273667.1 | 5 | 797 | |||
XM_005273611.4 | XP_005273668.1 | 5 | 700 | |||
XM_005273612.4 | XP_005273669.1 | 5 | 794 | |||
XM_017013746.1 | XP_016869235.1 | 4 | 735 | |||
XM_017013747.1 | XP_016869236.1 | 5 | 797 | |||
GenBank mRNA | AK314287.1 | - | 4 | 734 | ||
BC069648.1 | - | 4 | 639 | 7262473 | ||
BC104905.1 | - | 4 | 625 | 8143908 | ||
BC113661.1 | - | 4 | 625 | 8322713 | ||
BC143583.1 | - | 4 | 625 | 9052094 | ||
D29990.1 | - | 3 | 564 | |||
U76368.1 | - | 4 | 758 |