Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138508.4 | NP_612517.2 | 1, 2 | 85 | ||
GenBank mRNA | BC081713.1 | - | 1, 2 | 98 | 7109192 | |
M57453.1 | - | 1, 2 | 85 | |||
M62763.1 | - | 1, 2 | 112 | |||
X60654.1 | - | 1, 2 | 65 |