Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013031.1 | NP_037163.1 | 12 | 1202 | ||
XM_006231664.2 | XP_006231726.1 | 12 | 2043 | |||
XM_008760515.1 | XP_008758737.1 | 11 | 1402 | |||
XM_008760516.1 | XP_008758738.1 | 12 | 2048 | |||
XM_008760517.1 | XP_008758739.1 | 12 | 2043 | |||
XM_008760518.1 | XP_008758740.1 | 11 | 1402 | |||
XM_008760519.1 | XP_008758741.1 | 10 | 1309 | |||
XM_008760520.1 | XP_008758742.1 | 9 | 1225 | |||
GenBank mRNA | L00603.1 | - | 12 | 1202 | ||
M97381.1 | - | 12 | 1173 |