Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013032.3 | NP_037164.3 | 8 | 897 | ||
GenBank mRNA | BC061743.1 | - | 8 | 897 | 5599027 | |
D63772.1 | - | 928 | ||||
L35558.1 | - | 3 | 343 | |||
U21104.1 | - | 8 | 909 | |||
U39555.1 | - | 8 | 987 | |||
X94255.1 | - | 8 | 954 |