Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017102.2 | NP_058798.2 | 1063 | |||
XM_006237295.1 | XP_006237357.1 | 1291 | ||||
XM_006237296.2 | XP_006237358.1 | 1610 | ||||
XM_006237297.1 | XP_006237359.1 | 1295 | ||||
XM_006237298.2 | XP_006237360.1 | 1573 | ||||
GenBank mRNA | D13962.1 | - | 1147 | |||
U17978.1 | - | 1063 |
Gene Symbol: LOC100909595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749833.3 | XP_003749881.1 | 8, 9 | 1397 |