Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017102.2 | NP_058798.2 | 1294 | |||
XM_006237295.1 | XP_006237357.1 | 1522 | ||||
XM_006237296.2 | XP_006237358.1 | 1841 | ||||
XM_006237297.1 | XP_006237359.1 | 1526 | ||||
XM_006237298.2 | XP_006237360.1 | 1804 | ||||
GenBank mRNA | D13962.1 | - | 1378 | |||
U17978.1 | - | 1294 |
Gene Symbol: LOC100909595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749833.3 | XP_003749881.1 | 10 | 1628 |