Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013039.2 | NP_037171.2 | 7, 8 | 1173 | ||
XM_008759319.1 | XP_008757541.1 | 6, 7 | 1561 | |||
XM_008759320.1 | XP_008757542.1 | 7, 8 | 1245 | |||
XM_008759321.1 | XP_008757543.1 | 7, 8 | 1167 | |||
XM_008759322.1 | XP_008757544.1 | 7, 8 | 1245 | |||
GenBank mRNA | AB052294.1 | - | 7, 8 | 1188 | ||
AF039595.1 | - | 7, 8 | 1173 | |||
AJ511272.1 | - | 7, 8 | 1095 | |||
AJ511273.1 | - | 7, 8 | 1152 | |||
X97279.1 | - | 7, 8 | 1173 |