Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tff3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013042.2 | NP_037174.2 | 2 | 212 | ||
GenBank mRNA | AF012534.1 | - | 2 | 112 | ||
BC078873.1 | - | 2 | 192 | 7127312 | ||
BP490107.1 | - | 2 | 212 | |||
M80826.1 | - | 2 | 191 | |||
S49317.1 | - | 2 | 186 | |||
U48825.1 | - | 2 | 185 | |||
X66956.1 | - | 2 | 186 |