Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109912.1 | NP_001103382.1 | 1 | 2938 | ||
XM_006252339.1 | XP_006252401.1 | 2 | 2810 | |||
XM_006252340.2 | XP_006252402.1 | 1 | 2941 |