Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004173.2 | NP_004164.2 | 3 | 1669 | ||
GenBank mRNA | AJ000730.1 | - | 3 | 1682 | ||
BC008814.2 | - | 3 | 1669 | 3958091 | ||
BC062565.1 | - | 3 | 1663 | 5165369 | ||
BC107160.1 | - | 2 | 1632 | 40033862 | ||
BC107161.1 | - | 2 | 1632 | 40033864 | ||
CR456580.1 | - | 2 | 1634 |