Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033236.1 | NP_150239.1 | 8 | 751 | ||
GenBank mRNA | BC061542.1 | - | 8 | 793 | 6918933 | |
D50694.1 | - | 8 | 751 | |||
FQ212821.1 | - | 8 | 781 | |||
FQ213265.1 | - | 5 | 554 | |||
FQ215767.1 | - | 5 | 554 | |||
FQ225383.1 | - | 5 | 554 |