Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013059.1 | NP_037191.1 | 3 | 281 | ||
XM_006239136.2 | XP_006239198.1 | 3 | 655 | |||
GenBank mRNA | BC088399.1 | - | 3 | 304 | 7128677 | |
J03572.1 | - | 3 | 281 | |||
Y00714.1 | - | 2 | 131 |