Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013060.3 | NP_037192.1 | 1 | 379 | ||
GenBank mRNA | AY321351.1 | - | 512 | |||
BC086391.1 | - | 374 | 7303450 | |||
D10863.1 | - | 1 | 379 | |||
FQ212969.1 | - | 397 | ||||
FQ213471.1 | - | 397 | ||||
FQ213662.1 | - | 397 | ||||
FQ220574.1 | - | 1 | 397 | |||
FQ227397.1 | - | 1 | 397 |