Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013066.1 | NP_037198.1 | 10 | 5136 | ||
XM_006245100.2 | XP_006245162.1 | 10 | 9285 | |||
XM_008767197.1 | XP_008765419.1 | 10 | 5316 | |||
XM_008767198.1 | XP_008765420.1 | 10 | 5316 | |||
XM_008767199.1 | XP_008765421.1 | 10 | 5313 | |||
XM_008767200.1 | XP_008765422.1 | 10 | 5313 | |||
XM_008767201.1 | XP_008765423.1 | 10 | 5313 | |||
XM_008767202.1 | XP_008765424.1 | 10 | 5304 | |||
XM_008767203.1 | XP_008765425.1 | 10 | 5316 | |||
XM_008767204.1 | XP_008765426.1 | 9 | 5070 | |||
XM_008767205.1 | XP_008765427.1 | 9 | 5058 | |||
XM_008767206.1 | XP_008765428.1 | 10 | 5104 | |||
XM_008767207.1 | XP_008765429.1 | 8 | 1123 | |||
XM_008767208.1 | XP_008765430.1 | 8 | 1122 | |||
GenBank mRNA | BC081835.1 | - | 8 | 1134 | ||
X17682.1 | - | 7 | 1026 | |||
X51842.1 | - | 10 | 5108 | |||
X54100.1 | - | 10 | 5136 | |||
X74211.1 | - | 7 | 1009 |