Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013070.1 | NP_037202.1 | 23 | 3221 | ||
XM_006227637.2 | XP_006227699.1 | 22 | 3396 | |||
XM_008758593.1 | XP_008756815.1 | 23 | 3320 | |||
XM_008758594.1 | XP_008756816.1 | 23 | 3217 | |||
XM_008758595.1 | XP_008756817.1 | 22 | 3396 | |||
XM_008758596.1 | XP_008756818.1 | 22 | 3389 | |||
XM_008758597.1 | XP_008756819.1 | 22 | 3395 | |||
GenBank mRNA | AJ002967.1 | - | 23 | 3221 |