Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038597.2 | NP_001033686.1 | 2 | 634 | ||
NM_001038599.2 | NP_001033688.2 | 2 | 634 | |||
NM_001038600.2 | NP_001033689.1 | 2 | 634 | |||
NM_001038601.2 | NP_001033690.1 | 2 | 634 | |||
NM_013071.2 | NP_037203.1 | 2 | 634 | |||
XM_008758731.1 | XP_008756953.1 | 2 | 780 | |||
GenBank mRNA | AY225402.1 | - | 2 | 465 | ||
AY225403.1 | - | 2 | 465 | |||
AY309000.1 | - | 2 | 465 | |||
AY309002.1 | - | 2 | 459 | |||
AY309004.1 | - | 2 | 465 | |||
D16349.1 | - | 2 | 610 | |||
DQ680043.1 | - | 304 | ||||
EU024650.1 | - | 232 | ||||
EU024651.1 | - | 646 | ||||
EU024652.1 | - | 835 | ||||
EU340244.1 | - | 654 | ||||
EU340245.1 | - | 910 | ||||
EU340246.1 | - | 1096 | ||||
L13069.1 | - | 2 | 636 | |||
L20684.1 | - | 2 | 457 | |||
L22455.1 | - | 2 | 596 | |||
U02083.1 | - | 2 | 637 | |||
U35424.1 | - | 2 | 496 |
Set Membership: |
G-protein coupled receptor |