Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323972.1 | NP_001310901.1 | 13 | 1875 | ||
NM_001323973.1 | NP_001310902.1 | 14 | 2175 | |||
NM_001323974.1 | NP_001310903.1 | 14 | 2089 | |||
NM_001323975.1 | NP_001310904.1 | 14 | 2092 | |||
NM_004594.2 | NP_004585.1 | 14 | 2098 | |||
NR_136664.1 | 15 | 2170 | ||||
NR_136665.1 | 13 | 1968 | ||||
NR_136666.1 | 12 | 1929 | ||||
GenBank mRNA | AB209203.1 | - | 14 | 2981 | ||
AF111173.1 | - | 14 | 2104 | |||
AL137689.1 | - | 6 | 526 | |||
BC142671.1 | - | 14 | 2061 | 40148180 | ||
BC144131.1 | - | 13 | 1968 | |||
BC150207.1 | - | 13 | 2354 |