Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003049.3 | NP_003040.1 | 2 | 654 | ||
GenBank mRNA | AK314028.1 | - | 2 | 655 | ||
AY544127.1 | - | 2 | 581 | |||
BC069799.1 | - | 2 | 602 | 7262615 | ||
BC069822.1 | - | 2 | 601 | 7262603 | ||
BC074724.2 | - | 2 | 548 | 30915174 | ||
BC126298.1 | - | 2 | 574 | 8992014 | ||
BC136355.1 | - | 2 | 574 | 9020342 | ||
L21893.1 | - | 2 | 602 |