Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013112.1 | NP_037244.1 | 2 | 116 | ||
XM_006250238.2 | XP_006250300.1 | 3 | 250 | |||
XM_006250239.2 | XP_006250301.1 | 3 | 256 | |||
GenBank mRNA | FQ209753.1 | - | 3 | 181 | ||
FQ209821.1 | - | 3 | 181 | |||
FQ210214.1 | - | 3 | 174 | |||
FQ210245.1 | - | 3 | 181 | |||
FQ210295.1 | - | 3 | 178 | |||
FQ210325.1 | - | 3 | 201 | |||
FQ210458.1 | - | 3 | 178 | |||
FQ210462.1 | - | 3 | 179 | |||
FQ210537.1 | - | 2 | 175 | |||
FQ210542.1 | - | 2 | 304 | |||
FQ210648.1 | - | 3 | 178 | |||
FQ210885.1 | - | 3 | 208 | |||
FQ211006.1 | - | 3 | 175 | |||
FQ211265.1 | - | 3 | 178 | |||
FQ211383.1 | - | 3 | 180 | |||
FQ218273.1 | - | 3 | 181 | |||
FQ218326.1 | - | 3 | 175 | |||
FQ218756.1 | - | 3 | 182 | |||
FQ221741.1 | - | 3 | 181 | |||
FQ223965.1 | - | 3 | 175 | |||
M28615.1 | - | 2 | 85 | |||
X03468.1 | - | 2 | 116 |