Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000338.2 | NP_000329.2 | 6 | 986 | ||
NM_001184832.1 | NP_001171761.1 | 6 | 986 | |||
XM_005254606.1 | XP_005254663.1 | 6 | 986 | |||
GenBank mRNA | AK295375.1 | - | 5 | 439 | ||
AK298275.1 | - | 1 | 205 | |||
AK298312.1 | - | 6 | 985 | |||
BC040138.2 | - | 5 | 956 | |||
BX647067.1 | - | 1 | 4909 | |||
BX647484.1 | - | 2 | 4893 | |||
EF559316.2 | - | 5 | 784 | |||
U58130.1 | - | 5 | 789 |