Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013130.2 | NP_037262.2 | 5 | 1308 | ||
XM_006255394.1 | XP_006255456.1 | 5 | 1454 | |||
XM_006255395.2 | XP_006255457.1 | 5 | 2113 | |||
XM_006255396.2 | XP_006255458.1 | 5 | 1344 | |||
XM_006255397.1 | XP_006255459.1 | 5 | 1433 | |||
GenBank mRNA | AF067727.1 | - | 4 | 989 | ||
BC061757.1 | - | 5 | 1308 | 5599977 | ||
U66478.1 | - | 5 | 1289 |