Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013162.1 | NP_037294.1 | 5 | 426 | ||
GenBank mRNA | BC167099.1 | - | 5 | 427 | 7369314 | |
CO561550.1 | - | 5 | 426 | |||
DV725071.1 | - | 3 | 225 | |||
M10934.1 | - | 4 | 289 |