Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013168.2 | NP_037300.2 | 3 | 280 | ||
XM_006242896.1 | XP_006242958.1 | 3 | 366 | |||
XM_006242897.1 | XP_006242959.1 | 3 | 366 | |||
XM_008766148.1 | XP_008764370.1 | 3 | 366 | |||
XM_008766149.1 | XP_008764371.1 | 3 | 303 | |||
GenBank mRNA | BC088162.1 | - | 3 | 280 | 7375228 | |
FQ226062.1 | - | 3 | 203 | |||
FQ231091.1 | - | 3 | 264 | |||
FQ234870.1 | - | 2 | 352 | |||
X06827.1 | - | 3 | 155 | |||
Y12006.1 | - | 3 | 150 |